Gene Transcripts

Gene Symbol
LRRC8C
Entrez Gene ID
84230
Gene Name
leucine rich repeat containing 8 family member C
Gene Aliases
AD158, FAD158
Chromosome Location
Chr. 1: 89615810 - 89719535 on Build GRCh38
UniGene ID
Hs.412836
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032270.4 NP_115646.2 3 1108
XM_006710960.3 XP_006711023.1 3 2750
XM_011542282.2 XP_011540584.1 3 1095
XM_011542283.2 XP_011540585.1 4 2662
XM_017002503.1 XP_016857992.1 3 955
XM_017002504.1 XP_016857993.1 4 864
GenBank mRNA AB081509.1 - 3 989
AK127881.1 - 3 1045
AL136919.1 - 1 608
BC113973.1 - 3 915 40007964

View Full Product Details