Gene Transcripts

Gene Symbol
LZTS3
Entrez Gene ID
9762
Gene Name
leucine zipper tumor suppressor family member 3
Gene Aliases
PROSAPIP1
Chromosome Location
Chr. 20: 3162617 - 3173597 on Build GRCh38
UniGene ID
Hs.90232
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282533.1 NP_001269462.1 6 3561
XM_005260890.3 XP_005260947.1 5 3643
XM_005260892.1 XP_005260949.1 5 3699
XM_005260893.2 XP_005260950.1 5 3612
XM_011529407.2 XP_011527709.1 5 3580
XM_011529408.2 XP_011527710.1 5 4681
XM_011529409.2 XP_011527711.1 5 5101
GenBank mRNA AB011124.1 - 3 4727
BC038860.1 - 6 3517 5260571

View Full Product Details