Gene Transcripts
- Gene Symbol
- SCLT1
- Entrez Gene ID
- 132320
- Gene Name
- sodium channel and clathrin linker 1
- Gene Aliases
- CAP-1A, CAP1A
- Chromosome Location
- Chr. 4: 128875206 - 129093607 on Build GRCh38
- UniGene ID
- Hs.654690
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_144643.3 |
NP_653244.2
|
17, 18 |
2133 |
|
XM_017007716.1 |
XP_016863205.1
|
17, 18 |
2145 |
|
XM_017007717.1 |
XP_016863206.1
|
17, 18 |
2145 |
|
XM_017007718.1 |
XP_016863207.1
|
17, 18 |
2145 |
|
XM_017007719.1 |
XP_016863208.1
|
17, 18 |
1666 |
|
XM_017007720.1 |
XP_016863209.1
|
18, 19 |
2045 |
|
XM_017007721.1 |
XP_016863210.1
|
11, 12 |
1247 |
|
GenBank mRNA |
AK055217.1 |
- |
17, 18 |
2133 |
|
AK093361.1 |
- |
11, 12 |
1286 |
|
AK122852.1 |
- |
7, 8 |
2758 |
|
View Full Product Details