Gene Transcripts
- Gene Symbol
- SDK1
- Entrez Gene ID
- 221935
- Gene Name
- sidekick cell adhesion molecule 1
- Gene Aliases
- Chromosome Location
- Chr. 7: 3301448 - 4269000 on Build GRCh38
- UniGene ID
- Hs.653013
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_152744.3 |
NP_689957.3
|
4, 5 |
851 |
|
GenBank mRNA |
AY310398.1 |
- |
4, 5 |
851 |
|
View Full Product Details