Gene Transcripts
- Gene Symbol
- TDRD5
- Entrez Gene ID
- 163589
- Gene Name
- tudor domain containing 5
- Gene Aliases
- TUDOR3
- Chromosome Location
- Chr. 1: 179591613 - 179691272 on Build GRCh38
- UniGene ID
- Hs.197354
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001199085.1 |
NP_001186014.1
|
16, 17 |
3175 |
|
NM_001199089.1 |
NP_001186018.1
|
16, 17 |
2888 |
|
NM_001199091.1 |
NP_001186020.1
|
15, 16 |
2854 |
|
NM_001199092.1 |
NP_001186021.1
|
12, 13 |
1698 |
|
NM_173533.3 |
NP_775804.2
|
15, 16 |
3013 |
|
XM_005244934.1 |
XP_005244991.1
|
16, 17 |
2960 |
|
XM_005244935.4 |
XP_005244992.1
|
16, 17 |
2865 |
|
XM_017000473.1 |
XP_016855962.1
|
15, 16 |
2719 |
|
GenBank mRNA |
AK092142.1 |
- |
12, 13 |
1710 |
|
AK126889.1 |
- |
15, 16 |
2854 |
|
AK127370.1 |
- |
15, 16 |
2745 |
|
BC043368.1 |
- |
7, 8 |
1025 |
|
BC089387.1 |
- |
13, 14 |
1915 |
|
BC130532.1 |
- |
16, 17 |
2852 |
40146563 |
BC144058.1 |
- |
16, 17 |
2907 |
9052578 |
View Full Product Details