Gene Transcripts
- Gene Symbol
- SUMF1
- Entrez Gene ID
- 285362
- Gene Name
- sulfatase modifying factor 1
- Gene Aliases
- AAPA3037, FGE, UNQ3037
- Chromosome Location
- Chr. 3: 4034714 - 4467282 on Build GRCh38
- UniGene ID
- Hs.350475
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001164675.1 |
NP_001158147.1
|
3, 4 |
548 |
|
NM_182760.3 |
NP_877437.2
|
3, 4 |
548 |
|
XM_011533624.2 |
XP_011531926.1
|
3, 4 |
539 |
|
XM_011533625.2 |
XP_011531927.1
|
3, 4 |
539 |
|
XM_011533626.2 |
XP_011531928.1
|
3, 4 |
539 |
|
XM_017006252.1 |
XP_016861741.1
|
3, 4 |
539 |
|
XM_017006254.1 |
XP_016861743.1
|
3, 4 |
539 |
|
XM_017006255.1 |
XP_016861744.1
|
3, 4 |
539 |
|
GenBank mRNA |
AB448737.1 |
- |
3, 4 |
519 |
|
AK057983.1 |
- |
4, 5 |
447 |
|
AK075459.1 |
- |
3, 4 |
530 |
|
AY208752.1 |
- |
3, 4 |
511 |
|
AY323910.1 |
- |
3, 4 |
511 |
|
AY358092.1 |
- |
3, 4 |
528 |
|
BC110862.1 |
- |
3, 4 |
519 |
5760490 |
BC121122.2 |
- |
3, 4 |
540 |
40120670 |
BC121123.2 |
- |
3, 4 |
540 |
40120671 |
BG705404.1 |
- |
3, 4 |
541 |
|
View Full Product Details