Gene Transcripts

Gene Symbol
TTYH1
Entrez Gene ID
57348
Gene Name
tweety family member 1
Gene Aliases
Chromosome Location
Chr. 19: 54415431 - 54436719 on Build GRCh38
UniGene ID
Hs.268728
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005367.2 NP_001005367.1 13 1535
NM_001201461.1 NP_001188390.1 14 1584
NM_020659.3 NP_065710.1 14 1581
XM_017027025.1 XP_016882514.1 13 1379
XM_017027026.1 XP_016882515.1 14 1425
GenBank mRNA AB162930.1 - 13 1418
AB188496.1 - 14 1584
AF177909.1 - 14 1495
AK289468.1 - 13 1498
BC011347.1 - 13 1440 4215136
BC019358.1 - 11 917 3960435

View Full Product Details