Gene Transcripts

Gene Symbol
RAB22A
Entrez Gene ID
57403
Gene Name
RAB22A, member RAS oncogene family
Gene Aliases
Chromosome Location
Chr. 20: 58309715 - 58367507 on Build GRCh38
UniGene ID
Hs.529044
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020673.2 NP_065724.1 7 5858
GenBank mRNA AK054767.1 - 1 70

View Full Product Details