Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001287031.1 | NP_001273960.1 | 6 | 575 | |
NM_001287032.1 | NP_001273961.1 | 7 | 599 | ||
NM_001287033.1 | NP_001273962.1 | 6 | 572 | ||
NM_013442.2 | NP_038470.1 | 7 | 710 | ||
GenBank mRNA | AF190167.1 | - | 7 | 672 | |
AF282596.1 | - | 7 | 643 | ||
AK027405.1 | - | 7 | 643 | ||
AK130546.1 | - | 4 | 1490 | ||
AK223102.1 | - | 7 | 659 | ||
AK225114.1 | - | 7 | 643 | ||
AK303883.1 | - | 6 | 513 | ||
BC002442.2 | - | 7 | 646 | 3346384 | |
BC003025.1 | - | 7 | 631 | 2821269 | |
BC010152.2 | - | 7 | 643 | 3534656 | |
BC014990.1 | - | 7 | 641 | 4907745 | |
BQ935988.1 | - | 7 | 587 | ||
BU528995.1 | - | 6 | 469 | ||
BX400542.2 | - | 7 | 597 |