Gene Transcripts

Gene Symbol
RNF11
Entrez Gene ID
26994
Gene Name
ring finger protein 11
Gene Aliases
CGI-123, SID1669
Chromosome Location
Chr. 1: 51236273 - 51273447 on Build GRCh38
UniGene ID
Hs.309641
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014372.4 NP_055187.1 3 1164
GenBank mRNA AB024703.1 - 3 807
AF151881.1 - 3 807
AK293047.1 - 5 993
BC020964.2 - 3 759 3917224

View Full Product Details