Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_015397.3 | NP_056212.1 | 8 | 1477 | |
XM_011517843.1 | XP_011516145.1 | 8 | 1304 | ||
GenBank mRNA | AB067479.1 | - | 8 | 1397 | |
AF131771.1 | - | 3 | 337 | ||
AK000354.1 | - | 8 | 1255 | ||
AK292935.1 | - | 8 | 1283 | ||
AL117646.1 | - | 5 | 611 | ||
BC008893.2 | - | 7 | 1040 | ||
BC063823.1 | - | 8 | 1395 | 6184796 | |
DQ518909.1 | - | 8 | 1397 |