Gene Transcripts

Gene Symbol
SPRY3
Entrez Gene ID
10251
Gene Name
sprouty RTK signaling antagonist 3
Gene Aliases
spry-3
Chromosome Location
Chr. X: 155612565 - 155782459 on Build GRCh38
UniGene ID
Hs.381912
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001304990.1 NP_001291919.1 3 611
NM_005840.2 NP_005831.1 2 755
GenBank mRNA AK125483.1 - 2 680
AK289543.1 - 2 748
BC069609.1 - 1 317 7262284
BC074972.2 - 1 293 30915293
BC074973.2 - 1 293 30915511

View Full Product Details