Gene Transcripts
- Gene Symbol
- ZER1
- Entrez Gene ID
- 10444
- Gene Name
- zyg-11 related cell cycle regulator
- Gene Aliases
- C9orf60, ZYG, ZYG11BL
- Chromosome Location
- Chr. 9: 128729786 - 128772507 on Build GRCh38
- UniGene ID
- Hs.147950
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_006336.3 |
NP_006327.2
|
2, 3 |
600 |
|
XM_005251645.2 |
XP_005251702.1
|
2, 3 |
392 |
|
XM_005251646.2 |
XP_005251703.1
|
2, 3 |
595 |
|
XM_011518122.2 |
XP_011516424.1
|
2, 3 |
402 |
|
XM_017014185.1 |
XP_016869674.1
|
2, 3 |
1008 |
|
XM_017014186.1 |
XP_016869675.1
|
2, 3 |
389 |
|
XM_017014187.1 |
XP_016869676.1
|
2, 3 |
1008 |
|
XM_017014188.1 |
XP_016869677.1
|
2, 3 |
376 |
|
XM_017014189.1 |
XP_016869678.1
|
2, 3 |
595 |
|
XM_017014190.1 |
XP_016869679.1
|
2, 3 |
389 |
|
GenBank mRNA |
AK314911.1 |
- |
2, 3 |
348 |
|
BC033884.1 |
- |
2, 3 |
569 |
|
BC052563.1 |
- |
2, 3 |
585 |
6303965 |
X99802.1 |
- |
1, 2 |
190 |
|
View Full Product Details