Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001017524.2 | NP_001017524.1 | 7 | 1467 | |
NM_012239.5 | NP_036371.1 | 7 | 1613 | ||
XM_005252835.1 | XP_005252892.1 | 7 | 1667 | ||
XM_011519956.1 | XP_011518258.1 | 7 | 2459 | ||
XM_011519957.1 | XP_011518259.1 | 7 | 2288 | ||
XM_017017428.1 | XP_016872917.1 | 7 | 1513 | ||
XM_017017429.1 | XP_016872918.1 | 7 | 1687 | ||
XM_017017430.1 | XP_016872919.1 | 7 | 1627 | ||
XM_017017431.1 | XP_016872920.1 | 7 | 1412 | ||
GenBank mRNA | AF083108.2 | - | 7 | 1681 | |
AK074992.1 | - | 5 | 2453 | ||
AK298572.1 | - | 7 | 1362 | ||
AK301979.1 | - | 7 | 1655 | ||
AL137276.1 | - | 3 | 736 | ||
BC001042.2 | - | 7 | 1596 | 2823471 | |
DR762907.1 | - | 1 | 327 |