Gene Transcripts
- Gene Symbol
- RPH3A
- Entrez Gene ID
- 22895
- Gene Name
- rabphilin 3A
- Gene Aliases
- Chromosome Location
- Chr. 12: 112791538 - 112898881 on Build GRCh38
- UniGene ID
- Hs.21239
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001143854.1 |
NP_001137326.1
|
17, 18 |
1967 |
|
NM_014954.3 |
NP_055769.2
|
16, 17 |
1955 |
|
XM_011538048.1 |
XP_011536350.1
|
17, 18 |
1771 |
|
XM_017019034.1 |
XP_016874523.1
|
9, 10 |
1225 |
|
GenBank mRNA |
AB023202.1 |
- |
17, 18 |
1888 |
|
AK295696.1 |
- |
17, 18 |
1921 |
|
AK296296.1 |
- |
15, 16 |
1506 |
|
AK316112.1 |
- |
17, 18 |
1802 |
|
BC002605.2 |
- |
15, 16 |
1693 |
|
BC009843.2 |
- |
15, 16 |
1616 |
|
BC017259.2 |
- |
16, 17 |
1762 |
3510158 |
BX647281.1 |
- |
9, 10 |
2561 |
|
View Full Product Details