Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_015024.4 | NP_055839.3 | 22 | 2518 | |
XM_017013244.1 | XP_016868733.1 | 22 | 2772 | ||
XM_017013245.1 | XP_016868734.1 | 22 | 2539 | ||
XM_017013246.1 | XP_016868735.1 | 22 | 2741 | ||
GenBank mRNA | AB018288.1 | - | 19 | 1882 | |
AF064729.1 | - | 22 | 2428 | ||
AK291472.1 | - | 22 | 2433 | ||
AK299848.1 | - | 18 | 1954 | ||
BC030785.1 | - | 22 | 2517 | 5269230 |