Gene Transcripts
- Gene Symbol
- PSTPIP1
- Entrez Gene ID
- 9051
- Gene Name
- proline-serine-threonine phosphatase interacting protein 1
- Gene Aliases
- CD2BP1, CD2BP1L, CD2BP1S, H-PIP, PAPAS, PSTPIP
- Chromosome Location
- Chr. 15: 76994680 - 77037475 on Build GRCh38
- UniGene ID
- Hs.129758
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001321135.1 |
NP_001308064.1
|
5, 6 |
836 |
|
NM_001321136.1 |
NP_001308065.1
|
6, 7 |
1055 |
|
NM_001321137.1 |
NP_001308066.1
|
6, 7 |
652 |
|
NM_003978.4 |
NP_003969.2
|
5, 6 |
836 |
|
NR_135552.1 |
|
5, 6 |
836 |
|
GenBank mRNA |
AB451310.1 |
- |
5, 6 |
347 |
|
AB451440.1 |
- |
5, 6 |
347 |
|
AF038602.1 |
- |
5, 6 |
347 |
|
AF038603.1 |
- |
5, 6 |
347 |
|
AK308462.1 |
- |
6, 7 |
652 |
|
AK309856.1 |
- |
6, 7 |
1018 |
|
BC008602.1 |
- |
5, 6 |
708 |
4180398 |
KU178458.1 |
- |
5, 6 |
347 |
|
KU178459.1 |
- |
5, 6 |
347 |
|
U94778.1 |
- |
5, 6 |
563 |
|
View Full Product Details