Gene Transcripts

Gene Symbol
PRPH2
Entrez Gene ID
5961
Gene Name
peripherin 2
Gene Aliases
AOFMD, AVMD, CACD2, DS, MDBS1, PRPH, RDS, RP7, TSPAN22, rd2
Chromosome Location
Chr. 6: 42694509 - 42723024 on Build GRCh38
UniGene ID
Hs.654489
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000322.4 NP_000313.2 3 1245
GenBank mRNA BC074720.2 - 3 985
M62958.1 - 3 1207
M73531.1 - 3 1207

View Full Product Details