Gene Transcripts

Gene Symbol
KPNA3
Entrez Gene ID
3839
Gene Name
karyopherin subunit alpha 3
Gene Aliases
IPOA4, SRP1, SRP1gamma, SRP4, hSRP1
Chromosome Location
Chr. 13: 49699307 - 49792921 on Build GRCh38
UniGene ID
Hs.527919
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002267.3 NP_002258.2 17 3961
XM_017020561.1 XP_016876050.1 17 3644
GenBank mRNA BC035090.2 - 17 3963

View Full Product Details