Gene Transcripts

Gene Symbol
VPS13D
Entrez Gene ID
55187
Gene Name
vacuolar protein sorting 13 homolog D
Gene Aliases
Chromosome Location
Chr. 1: 12230039 - 12512047 on Build GRCh38
UniGene ID
Hs.439381
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015378.3 NP_056193.2 4 486
NM_018156.3 NP_060626.2 4 486
GenBank mRNA AJ608774.1 - 4 458
AJ608775.1 - 4 458
AL162331.1 - 4 458

View Full Product Details