Gene Transcripts

Gene Symbol
RAB5C
Entrez Gene ID
5878
Gene Name
RAB5C, member RAS oncogene family
Gene Aliases
L1880, RAB5CL, RAB5L, RABL
Chromosome Location
Chr. 17: 42124976 - 42155044 on Build GRCh38
UniGene ID
Hs.650382
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001252039.1 NP_001238968.1 5, 6 905
NM_004583.3 NP_004574.2 4, 5 689
NM_201434.2 NP_958842.1 5, 6 806
XM_011525090.1 XP_011523392.1 4, 5 628
XM_011525091.1 XP_011523393.1 4, 5 752
GenBank mRNA AF141304.1 - 3, 4 518
AF498938.1 - 3, 4 449
AK310234.1 - 5, 6 849
AL553867.3 - 4, 5 644
BC106039.1 - 4, 5 648 4938169
BC114439.1 - 4, 5 644 40083572
BM424166.1 - 5, 6 746
BM550607.1 - 4, 5 649
BT019484.1 - 3, 4 449
CR541901.1 - 3, 4 449
U11293.1 - 3, 4 524
U18420.1 - 4, 5 584

View Full Product Details