Gene Transcripts

Gene Symbol
FSD1L
Entrez Gene ID
83856
Gene Name
fibronectin type III and SPRY domain containing 1 like
Gene Aliases
CCDC10, CSDUFD1, FSD1CL, FSD1NL, MIR1
Chromosome Location
Chr. 9: 105442183 - 105552433 on Build GRCh38
UniGene ID
Hs.136901
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145313.2 NP_001138785.1 14 1929
NM_001287191.1 NP_001274120.1 14 1863
NM_001287192.1 NP_001274121.1 13 1830
XM_005252254.2 XP_005252311.1 15 2195
XM_005252257.3 XP_005252314.1 14 1861
XM_005252260.3 XP_005252317.1 13 1828
XM_011519077.2 XP_011517379.1 16 1853
XM_011519078.2 XP_011517380.2 14 1827
XM_011519079.2 XP_011517381.2 14 1826
XM_011519080.2 XP_011517382.1 9 2558
XM_011519081.2 XP_011517383.1 8 2510
XM_017015182.1 XP_016870671.1 14 2159
XM_017015183.1 XP_016870672.1 13 1828
XM_017015184.1 XP_016870673.1 10 1371
XM_017015185.1 XP_016870674.1 11 1393
XM_017015186.1 XP_016870675.1 9 1211
XM_017015187.1 XP_016870676.1 10 1386
GenBank mRNA AK296433.1 - 14 1774
BC036746.1 - 8 1272 4384472
CR936789.1 - 5 921

View Full Product Details