Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001102607.1 | NP_001096077.1 | 4 | 1011 | |
NM_172927.3 | NP_766515.2 | 4 | 1046 | ||
GenBank mRNA | AK054356.1 | - | 4 | 1047 | |
AK087828.1 | - | 4 | 1087 | ||
AM748260.1 | - | 2 | 731 | ||
BC150695.1 | - | 3 | 830 | 9087606 |