Gene Transcripts

Gene Symbol
Slc24a5
Entrez Gene ID
317750
Gene Name
solute carrier family 24, member 5
Gene Aliases
F630045L20Rik, JSX, NCKX5, NCX5, Oca6
Chromosome Location
Chr. 2: 125068127 - 125088677 on Build GRCm38
UniGene ID
Mm.330538
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175034.3 NP_778199.2 4 481
GenBank mRNA AB085629.1 - 4 484
AK089225.1 - 4 485
AY236527.1 - 4 451
BC094232.1 - 4 441 30538386
BC144866.1 - 4 476 9053414
BC145643.1 - 4 476 40130946

View Full Product Details