Gene Transcripts

Gene Symbol
Elk4
Entrez Gene ID
13714
Gene Name
ELK4, member of ETS oncogene family
Gene Aliases
2310011G17Rik, A130026I01Rik, BB162516, SAP-1, Sap1
Chromosome Location
Chr. 1: 132007234 - 132032612 on Build GRCm38
UniGene ID
Mm.254233
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007923.2 NP_031949.2 3 1320
GenBank mRNA AK156537.1 - 3 1317
BC004798.1 - 1 565
BC041679.1 - 1 553
BC144844.1 - 3 1070 9053392
BC145795.1 - 3 1070 40131098
Z36885.1 - 4 1265

View Full Product Details