Gene Transcripts

Gene Symbol
Drd2
Entrez Gene ID
13489
Gene Name
dopamine receptor D2
Gene Aliases
D2R, Drd-2
Chromosome Location
Chr. 9: 49340428 - 49408174 on Build GRCm38
UniGene ID
Mm.41970
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010077.2 NP_034207.2 8 1280
GenBank mRNA BC105663.1 - 8 1280
BC105664.1 - 8 1280 40046668
BC105665.1 - 8 1280 40046660
BC105666.1 - 8 1280 40046662
X55674.1 - 8 1315

View Full Product Details