Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_005558.3 | NP_005549.2 | 3 | 651 | |
GenBank mRNA | AK310917.1 | - | 4 | 537 | |
AK313388.1 | - | 3 | 434 | ||
BC009742.2 | - | 3 | 651 | 3842503 | |
BC071890.1 | - | 3 | 712 | 6082757 | |
BX647307.1 | - | 3 | 459 | ||
U42408.1 | - | 3 | 635 |