Gene Transcripts

Gene Symbol
Rwdd4a
Entrez Gene ID
192174
Gene Name
RWD domain containing 4A
Gene Aliases
BC016198, Gm1942, Rwdd4
Chromosome Location
Chr. 8: 47533645 - 47552837 on Build GRCm38
UniGene ID
Mm.273081
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_203507.3 NP_987103.1 8 2471
GenBank mRNA AK004597.1 - 8 2425
AK038266.1 - 1 1900
BC016198.1 - 8 2407 4503404

View Full Product Details