Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001039157.2 | NP_001034246.1 | 5 | 594 | |
NM_001039158.2 | NP_001034247.1 | 4 | 568 | ||
NM_001039159.2 | NP_001034248.1 | 4 | 431 | ||
NM_001284312.1 | NP_001271241.1 | 5 | 495 | ||
NM_001284314.1 | NP_001271243.1 | 4 | 431 | ||
NM_021402.6 | NP_067377.4 | 5 | 731 | ||
GenBank mRNA | AF296656.1 | - | 3 | 251 | |
AK004626.1 | - | 4 | 415 | ||
AK076265.1 | - | 5 | 696 | ||
AK214508.1 | - | 3 | 203 | ||
AW826087.1 | - | 4 | 252 | ||
BC065779.1 | - | 5 | 495 | 1429290 | |
U93242.1 | - | 3 | 251 |