Gene Transcripts

Gene Symbol
Mylk
Entrez Gene ID
107589
Gene Name
myosin, light polypeptide kinase
Gene Aliases
9530072E15Rik, A930019C19Rik, AW489456, KRP, MLCK108, MLCK210, Mlck, nmMlck, smMLCK
Chromosome Location
Chr. 16: 34745199 - 35002436 on Build GRCm38
UniGene ID
Mm.33360
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_139300.3 NP_647461.3 32 5957
GenBank mRNA AF314149.1 - 3 782
AY237726.1 - 32 5986
AY237727.2 - 17 3510
BC034209.1 - 7 1104
BC037721.1 - 17 2520
BC038902.1 - 3 730
BC045197.1 - 17 3440
BC058610.1 - 32 5966 30056142

View Full Product Details