Gene Transcripts

Gene Symbol
TRDN
Entrez Gene ID
10345
Gene Name
triadin
Gene Aliases
CPVT5, TDN, TRISK
Chromosome Location
Chr. 6: 123216339 - 123637093 on Build GRCh38
UniGene ID
Hs.144744
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006073.3 NP_006064.2 34, 35 2157
GenBank mRNA CR749591.1 - 23, 24 866
U18985.1 - 34, 35 1861

View Full Product Details