Gene Transcripts

Gene Symbol
Pcdhgc5
Entrez Gene ID
93708
Gene Name
protocadherin gamma subfamily C, 5
Gene Aliases
Chromosome Location
Chr. 18: 37819546 - 37841873 on Build GRCm38
UniGene ID
Mm.247203
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033583.3 NP_291061.1 1 955
GenBank mRNA AF464160.1 - 1 851
AF464163.1 - 1 955
AY013813.1 - 1 826
BC138677.1 - 1 899 8861698
BC138678.1 - 1 1013 8861699

View Full Product Details