Gene Transcripts
- Gene Symbol
- Hvcn1
- Entrez Gene ID
- 74096
- Gene Name
- hydrogen voltage-gated channel 1
- Gene Aliases
- 0610039P13Rik, AI450555, BTS, HV1, Vsop., mVSOP
- Chromosome Location
- Chr. 5: 122206807 - 122242297 on Build GRCm38
- UniGene ID
- Mm.28804
- Species
- Mus musculus
- Gene Symbol
- Tctn1
- Entrez Gene ID
- 654470
- Gene Name
- tectonic family member 1
- Gene Aliases
- G730031O11Rik, Tect1
- Chromosome Location
- Chr. 5: 122239495 - 122264489 on Build GRCm38
- UniGene ID
- Mm.28804
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
DV657549.1 |
- |
1 |
572 |
|
View Full Product Details