Gene Transcripts

Gene Symbol
Nmral1
Entrez Gene ID
67824
Gene Name
NmrA-like family domain containing 1
Gene Aliases
1110025F24Rik, AI256624
Chromosome Location
Chr. 16: 4711317 - 4719356 on Build GRCm38
UniGene ID
Mm.28451
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290761.1 NP_001277690.1 3 841
NM_001290762.1 NP_001277691.1 3 841
NM_001290763.1 NP_001277692.1 3 841
NM_026393.2 NP_080669.1 4 871
GenBank mRNA AK012340.1 - 3 597
AK078389.1 - 3 597
AK146521.1 - 596
AK195249.1 - 2 169
BC030039.1 - 4 574 1313824

View Full Product Details