Gene Transcripts

Gene Symbol
Smarcc2
Entrez Gene ID
68094
Gene Name
SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily c, member 2
Gene Aliases
5930405J04Rik
Chromosome Location
Chr. 10: 128459236 - 128490583 on Build GRCm38
UniGene ID
Mm.417338
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001114096.1 NP_001107568.1 2 305
NM_001114097.1 NP_001107569.1 2 305
NM_198160.2 NP_937803.1 2 305
GenBank mRNA AK089461.1 - 2 240
AK146971.1 - 2 234
AK170530.1 - 2 240
BC040363.1 - 2 220
BC058720.1 - 2 247 5705438
CB248220.1 - 2 305

View Full Product Details