Gene Transcripts

Gene Symbol
Sdhd
Entrez Gene ID
66925
Gene Name
succinate dehydrogenase complex, subunit D, integral membrane protein
Gene Aliases
3110001M13Rik, AVLL5809, C78570, PRO19626
Chromosome Location
Chr. 9: 50596340 - 50603849 on Build GRCm38
UniGene ID
Mm.10406
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025848.3 NP_080124.1 4 1173
GenBank mRNA AK013503.1 - 4 1138
AK013962.2 - 4 1141
AK135970.1 - 4 1133

View Full Product Details