Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_006602.3 | NP_006593.2 | 5, 6 | 1481 | |
XM_005260185.3 | XP_005260242.1 | 5, 6 | 1385 | ||
GenBank mRNA | AB012124.1 | - | 5, 6 | 1342 | |
AF070992.1 | - | 5, 6 | 1319 | ||
AJ271337.1 | - | 4, 5 | 1190 |