Gene Transcripts

Gene Symbol
Fbxo17
Entrez Gene ID
50760
Gene Name
F-box protein 17
Gene Aliases
Fbg4, Fbx17, Fbxo26
Chromosome Location
Chr. 7: 28716748 - 28738144 on Build GRCm38
UniGene ID
Mm.358659
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015796.2 NP_056611.2 6 1108
GenBank mRNA AF176532.1 - 6 1025
AK133408.1 - 6 1068
BC026470.1 - 6 1094 4486850

View Full Product Details