Gene Transcripts

Gene Symbol
Ppm1d
Entrez Gene ID
53892
Gene Name
protein phosphatase 1D magnesium-dependent, delta isoform
Gene Aliases
AV338790, Wip1
Chromosome Location
Chr. 11: 85311254 - 85347071 on Build GRCm38
UniGene ID
Mm.45609
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016910.3 NP_058606.3 6 1761
GenBank mRNA AF200464.1 - 6 1542
AK005658.1 - 2 506
AK157818.1 - 6 1749
AK166632.1 - 6 967
AK171714.1 - 6 1741
BC023492.1 - 6 1637 5026376
BC051966.1 - 6 1761 5689560

View Full Product Details