Gene Transcripts

Gene Symbol
Svs2
Entrez Gene ID
53878
Gene Name
seminal vesicle secretory protein 2
Gene Aliases
BB115391, Semg1, SvsII
Chromosome Location
Chr. 2: 164235929 - 164238341 on Build GRCm38
UniGene ID
Mm.99395
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017390.4 NP_059086.3 2 353
GenBank mRNA AK035204.1 - 2 278
AK040724.1 - 2 279
AK090151.1 - 2 275
AK090302.1 - 2 279
AK136679.1 - 2 277
AK144285.1 - 2 278
AK144389.1 - 2 278
AK144429.1 - 2 278
AK144524.1 - 2 275
AK165704.1 - 2 278
AK165713.1 - 2 278
AK165727.1 - 2 275
AK165753.1 - 2 278
AK165825.1 - 2 278
BC107275.1 - 2 278 40052650
BC107276.1 - 2 278
DV056729.1 - 2 387

View Full Product Details