Gene Transcripts

Gene Symbol
Shoc2
Entrez Gene ID
56392
Gene Name
soc-2 (suppressor of clear) homolog (C. elegans)
Gene Aliases
AU017287, Sur8, mKIAA0862
Chromosome Location
Chr. 19: 53944232 - 54033280 on Build GRCm38
UniGene ID
Mm.228669
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001168505.1 NP_001161977.1 2 629
NM_019658.6 NP_062632.2 2 670
GenBank mRNA AF068921.1 - 1 305
AK146447.1 - 2 667
BC026364.1 - 1 536 4014650
BC049775.1 - 1 535 6515905
BC083060.1 - 2 629 5356514

View Full Product Details