Gene Transcripts

Gene Symbol
Rsrp1
Entrez Gene ID
27981
Gene Name
arginine/serine rich protein 1
Gene Aliases
2700043I21Rik, D4Ucla2, D4Wsu53e
Chromosome Location
Chr. 4: 134923625 - 134927370 on Build GRCm38
UniGene ID
Mm.331964
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK003799.1 - 3 894
AK028145.1 - 3 895
AK029529.1 - 3 893
AK160993.1 - 3 894
AK169760.1 - 3 894
BC043057.1 - 3 878 6419597
BC046231.1 - 3 795 6813388
BC054728.1 - 3 866 6419723
BC056986.1 - 3 827 6816649

View Full Product Details