Gene Transcripts

Gene Symbol
Wbp5
Entrez Gene ID
22381
Gene Name
WW domain binding protein 5
Gene Aliases
AL024233
Chromosome Location
Chr. X: 136245080 - 136247139 on Build GRCm38
UniGene ID
Mm.512
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011712.2 NP_035842.1 3 531
GenBank mRNA AK002214.1 - 3 485
AK213578.1 - 1 138
AY459189.1 - 3 530
BC007478.1 - 3 428 3490650
BC020289.1 - 3 456
BU511227.1 - 3 562
U92454.1 - 1 426

View Full Product Details