Gene Transcripts
- Gene Symbol
- STXBP2
- Entrez Gene ID
- 6813
- Gene Name
- syntaxin binding protein 2
- Gene Aliases
- FHL5, Hunc18b, MUNC18-2, UNC18-2, UNC18B, pp10122
- Chromosome Location
- Chr. 19: 7637101 - 7647874 on Build GRCh38
- UniGene ID
- Hs.515104
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001127396.2 |
NP_001120868.1
|
12, 13 |
1062 |
|
NM_001272034.1 |
NP_001258963.1
|
12, 13 |
1104 |
|
NM_006949.3 |
NP_008880.2
|
12, 13 |
1071 |
|
XM_011528210.1 |
XP_011526512.1
|
12, 13 |
1067 |
|
XM_011528212.2 |
XP_011526514.1
|
12, 13 |
1067 |
|
GenBank mRNA |
AB002559.1 |
- |
12, 13 |
1046 |
|
AK222967.1 |
- |
12, 13 |
1048 |
|
AK222977.1 |
- |
12, 13 |
1048 |
|
AK302257.1 |
- |
12, 13 |
924 |
|
AK303701.1 |
- |
12, 13 |
1092 |
|
BC002869.2 |
- |
12, 13 |
1036 |
3942156 |
BT006915.1 |
- |
12, 13 |
1013 |
|
U63533.1 |
- |
12, 13 |
1046 |
|
View Full Product Details