Gene Transcripts

Gene Symbol
Tas2r107
Entrez Gene ID
78981
Gene Name
taste receptor, type 2, member 107
Gene Aliases
T2R4, Tas2r10
Chromosome Location
Chr. 4: 165773200 - 165774126 on Build Rnor_6.0
UniGene ID
Rn.48784
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023995.1 NP_076485.1 1 533
GenBank mRNA AF227142.1 - 1 533

View Full Product Details