Gene Transcripts

Gene Symbol
Mia
Entrez Gene ID
81510
Gene Name
melanoma inhibitory activity
Gene Aliases
Cdrap, Mia1
Chromosome Location
Chr. 1: 83991577 - 83993270 on Build Rnor_6.0
UniGene ID
Rn.10660
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030852.2 NP_110479.1 3 363
GenBank mRNA U51438.1 - 3 253
U67884.1 - 3 363

View Full Product Details