Gene Transcripts
- Gene Symbol
- Mia
- Entrez Gene ID
- 81510
- Gene Name
- melanoma inhibitory activity
- Gene Aliases
- Cdrap, Mia1
- Chromosome Location
- Chr. 1: 83991577 - 83993270 on Build Rnor_6.0
- UniGene ID
- Rn.10660
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_030852.2 |
NP_110479.1
|
4 |
414 |
|
GenBank mRNA |
U67884.1 |
- |
4 |
414 |
|
View Full Product Details