Gene Transcripts

Gene Symbol
Vom1r90
Entrez Gene ID
266771
Gene Name
vomeronasal 1 receptor 90
Gene Aliases
V1ra16
Chromosome Location
Chr. 4: 121675261 - 121684264 on Build Rnor_6.0
UniGene ID
Rn.234590
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153729.2 NP_714951.1 3 383
GenBank mRNA AY510354.1 - 1 155
U36898.1 - 3 403

View Full Product Details