Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001321196.1 | NP_001308125.1 | 8 | 1603 | |
NM_012254.2 | NP_036386.1 | 8 | 1855 | ||
XM_011526364.1 | XP_011524666.1 | 8 | 1784 | ||
GenBank mRNA | AF064255.1 | - | 8 | 1815 | |
AK026640.1 | - | 1 | 121 | ||
AK123036.1 | - | 8 | 1855 | ||
AK298446.1 | - | 8 | 1530 |