Gene Transcripts

Gene Symbol
Mcpt9
Entrez Gene ID
54272
Gene Name
mast cell protease 9
Gene Aliases
Chromosome Location
Chr. 15: 35018087 - 35020842 on Build Rnor_6.0
UniGene ID
Rn.10651
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019323.1 NP_062196.1 293
GenBank mRNA U67912.1 - 274
U72143.1 - 293

View Full Product Details